Assay Details
| Gene Symbol: MAGEA12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005367.6 | NP_005358.2 | 1 - 2 | 78 | 70 | |
| GenBank mRNA | BC003408.2 | - | 1 - 2 | 66 | 3450217 | 70 |
| BQ436888.1 | - | 1 - 2 | 78 | 70 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X001503.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC003408.2,NM_005367.6 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019512.hg.1 | TC0X001503.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019512.hg.1 | ENST00000357916.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019512.hg.1 | ENST00000361201.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019512.hg.1 | ENST00000393869.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019512.hg.1 | NM_001166387.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019512.hg.1 | NM_005367.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019509.hg.1 | TC0X001503.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019509.hg.1 | ENST00000357916.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019509.hg.1 | ENST00000393869.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019509.hg.1 | ENST00000393900.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019509.hg.1 | NM_001166386.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019509.hg.1 | NM_001166387.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019509.hg.1 | NM_005367.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000357916.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000357916.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000357916.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000357916.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000357916.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000361201.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000361201.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000361201.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000361201.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000361201.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000393869.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000393869.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000393869.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000393869.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000393869.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000393869.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000393900.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000393900.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000393900.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000393900.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000393900.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000583763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000583763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | NM_001166386.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | NM_001166386.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | NM_001166386.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | NM_001166386.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | NM_001166386.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | NM_001166387.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | NM_001166387.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | NM_001166387.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | NM_001166387.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | NM_001166387.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | NM_001166387.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | NM_005367.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | NM_005367.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | NM_005367.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | NM_005367.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | NM_005367.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | NR_073432.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | NR_073432.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | NR_073432.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | NR_073432.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | NR_073432.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019508.hg.1 | TC0X001503.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019508.hg.1 | ENST00000357916.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019508.hg.1 | ENST00000393869.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019508.hg.1 | ENST00000393900.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019508.hg.1 | NM_001166386.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019508.hg.1 | NM_001166387.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019508.hg.1 | NM_005367.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019513.hg.1 | TC0X001503.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019513.hg.1 | ENST00000357916.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019513.hg.1 | ENST00000393869.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019513.hg.1 | NM_001166387.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019513.hg.1 | NM_005367.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159548.hg.1 | ENST00000357916.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159548.hg.1 | ENST00000393869.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159548.hg.1 | ENST00000393900.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159548.hg.1 | NM_001166386.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159548.hg.1 | NM_001166387.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159548.hg.1 | NM_005367.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159548.hg.1 | ENST00000357916.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159548.hg.1 | ENST00000393869.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159548.hg.1 | ENST00000393900.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159548.hg.1 | NM_001166386.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159548.hg.1 | NM_001166387.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159548.hg.1 | NM_005367.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159548.hg.1 | TC0X00008752.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000357916.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000357916.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000357916.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000357916.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000357916.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000361201.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000361201.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000361201.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000361201.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000361201.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000393869.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000393869.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000393869.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000393869.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000393869.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000393869.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000393900.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000393900.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000393900.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000393900.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000393900.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000583763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000583763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | NM_001166386.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | NM_001166386.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | NM_001166386.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | NM_001166386.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | NM_001166386.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | NM_001166387.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | NM_001166387.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | NM_001166387.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | NM_001166387.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | NM_001166387.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | NM_001166387.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | NM_005367.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | NM_005367.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | NM_005367.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | NM_005367.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | NM_005367.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | NR_073432.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | NR_073432.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | NR_073432.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | NR_073432.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | NR_073432.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159545.hg.1 | ENST00000357916.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159545.hg.1 | ENST00000361201.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159545.hg.1 | ENST00000393869.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159545.hg.1 | NM_001166387.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159545.hg.1 | NM_005367.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159545.hg.1 | NR_073432.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159545.hg.1 | ENST00000357916.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159545.hg.1 | ENST00000361201.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159545.hg.1 | ENST00000393869.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159545.hg.1 | NM_001166387.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159545.hg.1 | NM_005367.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159545.hg.1 | NR_073432.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159545.hg.1 | TC0X00008752.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00159544.hg.1 | ENST00000357916.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159544.hg.1 | ENST00000393869.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159544.hg.1 | NM_001166387.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159544.hg.1 | NM_005367.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159544.hg.1 | ENST00000357916.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159544.hg.1 | ENST00000393869.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159544.hg.1 | NM_001166387.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159544.hg.1 | NM_005367.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159544.hg.1 | TC0X00008752.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00159549.hg.1 | ENST00000357916.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159549.hg.1 | ENST00000393869.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159549.hg.1 | ENST00000393900.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159549.hg.1 | NM_001166386.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159549.hg.1 | NM_001166387.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159549.hg.1 | NM_005367.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159549.hg.1 | ENST00000357916.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159549.hg.1 | ENST00000393869.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159549.hg.1 | ENST00000393900.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159549.hg.1 | NM_001166386.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159549.hg.1 | NM_001166387.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159549.hg.1 | NM_005367.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159549.hg.1 | TC0X00008752.hg.1 | Clariom D TC |

MAGEA12
Genomic Map