Assay Details
| Gene Symbol: DHX40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166301.1 | NP_001159773.1 | 13 - 14 | 1724 | 90 | |
| NM_024612.4 | NP_078888.4 | 14 - 15 | 1955 | 90 | ||
| XM_006722083.2 | XP_006722146.1 | 13 - 14 | 1820 | 90 | ||
| XM_011525253.2 | XP_011523555.1 | 13 - 14 | 1718 | 90 | ||
| XM_011525254.2 | XP_011523556.1 | 14 - 15 | 1884 | 90 | ||
| XM_017025080.1 | XP_016880569.1 | 13 - 14 | 1719 | 90 | ||
| XM_017025081.1 | XP_016880570.1 | 13 - 14 | 1803 | 90 | ||
| XM_017025082.1 | XP_016880571.1 | 13 - 14 | 1771 | 90 | ||
| XM_017025083.1 | XP_016880572.1 | 13 - 14 | 1768 | 90 | ||
| GenBank mRNA | AF319521.1 | - | 832 | 90 | ||
| AF461690.1 | - | 14 - 15 | 1955 | 90 | ||
| AK025713.1 | - | 4 - 5 | 427 | 90 | ||
| AK091334.1 | - | 10 - 11 | 1247 | 90 | ||
| AK095681.1 | - | 14 - 15 | 1868 | 90 | ||
| AK225268.1 | - | 4 - 5 | 431 | 90 | ||
| AK225344.1 | - | 4 - 5 | 427 | 90 | ||
| AK226048.1 | - | 14 - 15 | 1833 | 90 | ||
| AK299151.1 | - | 13 - 14 | 1637 | 90 | ||
| AL832510.1 | - | 10 - 11 | 1173 | 90 | ||
| BC024187.2 | - | 14 - 15 | 1870 | 4822080 | 90 | |
More Information
Set Membership: |
|

DHX40
Genomic Map