Assay Details
| Gene Symbol: ABCA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005502.3 | NP_005493.2 | 27 - 28 | 4305 | 65 | |
| XM_005251773.2 | XP_005251830.1 | 27 - 28 | 4220 | 65 | ||
| XM_005251776.2 | XP_005251833.1 | 26 - 27 | 4060 | 65 | ||
| XM_011518339.2 | XP_011516641.1 | 28 - 29 | 4295 | 65 | ||
| XM_011518340.2 | XP_011516642.1 | 29 - 30 | 4510 | 65 | ||
| XM_011518341.2 | XP_011516643.1 | 28 - 29 | 4289 | 65 | ||
| XM_011518342.2 | XP_011516644.1 | 25 - 26 | 3998 | 65 | ||
| XM_011518344.2 | XP_011516646.1 | 28 - 29 | 4295 | 65 | ||
| XM_017014378.1 | XP_016869867.1 | 28 - 29 | 4493 | 65 | ||
| XM_017014379.1 | XP_016869868.1 | 28 - 29 | 4901 | 65 | ||
| XM_017014380.1 | XP_016869869.1 | 28 - 29 | 4227 | 65 | ||
| XM_017014381.1 | XP_016869870.1 | 28 - 29 | 4293 | 65 | ||
| XM_017014382.1 | XP_016869871.1 | 26 - 27 | 4758 | 65 | ||
| GenBank mRNA | AB055982.1 | - | 26 - 27 | 3901 | 65 | |
| AB445477.1 | - | 26 - 27 | 3901 | 65 | ||
| AF165281.1 | - | 25 - 26 | 3841 | 65 | ||
| AF285167.1 | - | 27 - 28 | 4191 | 65 | ||
| AJ012376.1 | - | 25 - 26 | 3841 | 65 | ||
| BC146856.1 | - | 27 - 28 | 4115 | 9056784 | 65 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001434.hg.1
Set Membership: |
|

ABCA1
Genomic Map