Assay Details
| Gene Symbol: CCR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005508.4 | NP_005499.1 | 2 | 1601 | 134 | |
| XM_017005687.1 | XP_016861176.1 | 2 | 1648 | 134 | ||
| GenBank mRNA | BC069139.1 | - | 1 | 1486 | 7216998 | 134 |
| BC071751.1 | - | 2 | 1601 | 30415446 | 134 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

CCR4
Genomic Map