Assay Details
| Gene Symbol: HSPB11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001316935.1 | NP_001303864.1 | 2 - 3 | 187 | 95 | |
| NM_016126.3 | NP_057210.2 | 2 - 3 | 384 | 95 | ||
| NR_133633.1 | 2 - 3 | 384 | 95 | |||
| GenBank mRNA | AF100747.1 | - | 2 - 3 | 157 | 95 | |
| AF416725.1 | - | 1 - 2 | 111 | 95 | ||
| AF417114.1 | - | 1 - 2 | 111 | 95 | ||
| BC005245.1 | - | 2 - 3 | 176 | 3838843 | 95 | |
| DA133366.1 | - | 2 - 3 | 260 | 95 | ||
| DB480705.1 | - | 2 - 3 | 384 | 95 | ||
| HY179788.1 | - | 2 - 3 | 188 | 95 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002679.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | ENST00000194214.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | ENST00000194214.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | ENST00000194214.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | ENST00000194214.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | ENST00000194214.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | ENST00000194214.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | ENST00000194214.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | ENST00000194214.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | ENST00000194214.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | ENST00000194214.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | ENST00000371376.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | ENST00000371376.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | ENST00000371376.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | ENST00000371376.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | ENST00000371377.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | ENST00000371377.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | ENST00000371377.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | ENST00000371377.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | ENST00000371377.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | ENST00000371377.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | ENST00000371377.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | ENST00000371378.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | ENST00000371378.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | ENST00000371378.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | ENST00000371378.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | ENST00000371378.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | ENST00000371378.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | ENST00000488884.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | ENST00000488884.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | ENST00000488884.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | ENST00000489675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | ENST00000489675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | ENST00000489675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | ENST00000489675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | ENST00000489675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | ENST00000489675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | NM_016126.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | NM_016126.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | NM_016126.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | NM_016126.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | NM_016126.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | NM_016126.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | NM_016126.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | NM_016126.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014247.hg.1 | NM_016126.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01042050.hg.1 | TC01002679.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01042050.hg.1 | ENST00000194214.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01042050.hg.1 | ENST00000371376.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01042050.hg.1 | ENST00000371377.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01042050.hg.1 | ENST00000371378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01042050.hg.1 | NM_016126.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01042046.hg.1 | TC01002679.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01042046.hg.1 | ENST00000194214.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01042046.hg.1 | ENST00000371377.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01042046.hg.1 | ENST00000371378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01042046.hg.1 | ENST00000489675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01042046.hg.1 | NM_016126.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01042043.hg.1 | TC01002679.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01042043.hg.1 | ENST00000194214.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01042043.hg.1 | ENST00000371378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01042043.hg.1 | ENST00000489675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01042043.hg.1 | NM_016126.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01042047.hg.1 | TC01002679.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01042047.hg.1 | ENST00000194214.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01042047.hg.1 | ENST00000371377.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01042047.hg.1 | ENST00000371378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01042047.hg.1 | ENST00000489675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01042047.hg.1 | NM_016126.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01042052.hg.1 | TC01002679.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01042052.hg.1 | ENST00000194214.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01042052.hg.1 | ENST00000371376.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01042052.hg.1 | ENST00000371377.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01042052.hg.1 | ENST00000371378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01042052.hg.1 | NM_016126.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01042054.hg.1 | TC01002679.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01042054.hg.1 | ENST00000194214.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01042054.hg.1 | ENST00000371377.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01042054.hg.1 | NM_016126.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | ENST00000194214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | ENST00000194214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | ENST00000194214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | ENST00000194214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | ENST00000194214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | ENST00000194214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | ENST00000194214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | ENST00000194214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | ENST00000194214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | ENST00000194214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | ENST00000371376.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | ENST00000371376.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | ENST00000371376.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | ENST00000371376.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | ENST00000371377.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | ENST00000371377.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | ENST00000371377.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | ENST00000371377.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | ENST00000371377.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | ENST00000371377.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | ENST00000371377.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | ENST00000371378.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | ENST00000371378.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | ENST00000371378.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | ENST00000371378.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | ENST00000371378.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | ENST00000371378.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | ENST00000488884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | ENST00000488884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | ENST00000488884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | ENST00000489675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | ENST00000489675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | ENST00000489675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | ENST00000489675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | ENST00000489675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | ENST00000489675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | NM_016126.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | NM_016126.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | NM_016126.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | NM_016126.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | NM_016126.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | NM_016126.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | NM_016126.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | NM_016126.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014247.hg.1 | NM_016126.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206708.hg.1 | ENST00000194214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206708.hg.1 | ENST00000371376.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206708.hg.1 | ENST00000371377.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206708.hg.1 | ENST00000371378.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206708.hg.1 | ENST00000488884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206708.hg.1 | NM_016126.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206708.hg.1 | ENST00000194214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206708.hg.1 | ENST00000371376.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206708.hg.1 | ENST00000371377.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206708.hg.1 | ENST00000371378.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206708.hg.1 | ENST00000488884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206708.hg.1 | NM_016126.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206708.hg.1 | TC0100014247.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100206712.hg.1 | ENST00000194214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206712.hg.1 | ENST00000371377.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206712.hg.1 | ENST00000488884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206712.hg.1 | NM_016126.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206712.hg.1 | ENST00000194214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206712.hg.1 | ENST00000371377.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206712.hg.1 | ENST00000488884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206712.hg.1 | NM_016126.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206712.hg.1 | TC0100014247.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100206706.hg.1 | ENST00000194214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206706.hg.1 | ENST00000371376.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206706.hg.1 | ENST00000371377.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206706.hg.1 | ENST00000371378.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206706.hg.1 | NM_016126.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206706.hg.1 | ENST00000194214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206706.hg.1 | ENST00000371376.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206706.hg.1 | ENST00000371377.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206706.hg.1 | ENST00000371378.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206706.hg.1 | NM_016126.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206706.hg.1 | TC0100014247.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100206705.hg.1 | ENST00000194214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206705.hg.1 | ENST00000371376.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206705.hg.1 | ENST00000371377.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206705.hg.1 | ENST00000371378.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206705.hg.1 | NM_016126.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206705.hg.1 | ENST00000194214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206705.hg.1 | ENST00000371376.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206705.hg.1 | ENST00000371377.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206705.hg.1 | ENST00000371378.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206705.hg.1 | NM_016126.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206705.hg.1 | TC0100014247.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 183023 | - | - |

HSPB11
Genomic Map