Assay Details
| Gene Symbol: SV2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014849.4 | NP_055664.3 | 10 - 11 | 2172 | 74 | |
| XM_017002992.1 | XP_016858481.1 | 10 - 11 | 1958 | 74 | ||
| GenBank mRNA | AB018279.1 | - | 10 - 11 | 2115 | 74 | |
| AK075480.1 | - | 10 - 11 | 2132 | 74 | ||
| BC000776.2 | - | 9 - 10 | 1267 | 74 | ||
| BC045111.1 | - | 10 - 11 | 2118 | 5804662 | 74 | |
| BX537754.1 | - | 10 - 11 | 2168 | 74 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003195.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | ENST00000369145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | ENST00000369145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | ENST00000369145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | ENST00000369145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | ENST00000369145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | ENST00000369145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | ENST00000369145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | ENST00000369145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | ENST00000369145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | ENST00000369145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | ENST00000369145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | ENST00000369145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | ENST00000369145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | ENST00000369145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | ENST00000369146.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | ENST00000369146.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | ENST00000369146.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | ENST00000369146.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | ENST00000369146.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | ENST00000369146.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | ENST00000369146.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | ENST00000369146.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | ENST00000369146.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | ENST00000369146.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | ENST00000369146.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | ENST00000369146.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | ENST00000369146.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | ENST00000369146.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | ENST00000369146.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | ENST00000369146.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | ENST00000369146.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | NM_001278719.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | NM_001278719.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | NM_001278719.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | NM_001278719.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | NM_001278719.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | NM_001278719.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | NM_014849.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | NM_014849.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | NM_014849.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | NM_014849.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | NM_014849.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | NM_014849.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | NM_014849.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | NM_014849.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | NM_014849.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | NM_014849.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | NM_014849.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | NM_014849.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | NM_014849.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | NM_014849.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | NM_014849.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | NM_014849.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015714.hg.1 | NM_014849.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01048940.hg.1 | TC01003195.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01048940.hg.1 | ENST00000369145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01048940.hg.1 | ENST00000369146.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01048940.hg.1 | NM_014849.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100217629.hg.1 | ENST00000369145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100217629.hg.1 | ENST00000369146.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100217629.hg.1 | NM_001278719.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100217629.hg.1 | NM_014849.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100217629.hg.1 | ENST00000369145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100217629.hg.1 | ENST00000369146.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100217629.hg.1 | NM_001278719.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100217629.hg.1 | NM_014849.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100217629.hg.1 | TC0100015714.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | ENST00000369145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | ENST00000369145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | ENST00000369145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | ENST00000369145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | ENST00000369145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | ENST00000369145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | ENST00000369145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | ENST00000369145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | ENST00000369145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | ENST00000369145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | ENST00000369145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | ENST00000369145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | ENST00000369145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | ENST00000369145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | ENST00000369146.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | ENST00000369146.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | ENST00000369146.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | ENST00000369146.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | ENST00000369146.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | ENST00000369146.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | ENST00000369146.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | ENST00000369146.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | ENST00000369146.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | ENST00000369146.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | ENST00000369146.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | ENST00000369146.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | ENST00000369146.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | ENST00000369146.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | ENST00000369146.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | ENST00000369146.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | ENST00000369146.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | NM_001278719.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | NM_001278719.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | NM_001278719.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | NM_001278719.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | NM_001278719.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | NM_001278719.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | NM_014849.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | NM_014849.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | NM_014849.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | NM_014849.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | NM_014849.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | NM_014849.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | NM_014849.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | NM_014849.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | NM_014849.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | NM_014849.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | NM_014849.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | NM_014849.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | NM_014849.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | NM_014849.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | NM_014849.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | NM_014849.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015714.hg.1 | NM_014849.4 | Refseq |

SV2A
Genomic Map