Assay Details
| Gene Symbol: TBATA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318241.1 | NP_001305170.1 | 9 - 10 | 1331 | 106 | |
| NM_001318242.1 | NP_001305171.1 | 9 - 10 | 1395 | 106 | ||
| NM_001318243.1 | NP_001305172.1 | 9 - 10 | 1325 | 106 | ||
| NM_152710.3 | NP_689923.3 | 9 - 10 | 1328 | 106 | ||
| NR_134531.1 | 8 - 9 | 1246 | 106 | |||
| NR_134532.1 | 9 - 10 | 1254 | 106 | |||
| NR_134533.1 | 5 - 6 | 864 | 106 | |||
| NR_134534.1 | 4 - 5 | 788 | 106 | |||
| XM_017015853.1 | XP_016871342.1 | 10 - 11 | 1406 | 106 | ||
| GenBank mRNA | AK057382.1 | - | 9 - 10 | 1310 | 106 | |
| BC137543.1 | - | 9 - 10 | 1362 | 9021543 | 106 | |
| BC137544.1 | - | 9 - 10 | 1296 | 9021544 | 106 | |
| BC139728.1 | - | 8 - 9 | 1195 | 40036949 | 106 | |
| BC144692.1 | - | 9 - 10 | 1293 | 9053226 | 106 | |
| BC144694.1 | - | 9 - 10 | 1299 | 9053228 | 106 | |
| HY289269.1 | - | 2 - 3 | 106 | |||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001367.hg.1
Set Membership: |
|

TBATA
Genomic Map