Assay Details
| Gene Symbol: NBR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291571.1 | NP_001278500.1 | 4 - 5 | 529 | 133 | |
| NM_005899.4 | NP_005890.2 | 4 - 5 | 529 | 133 | ||
| NM_031862.3 | NP_114068.1 | 4 - 5 | 337 | 133 | ||
| XM_006721903.1 | XP_006721966.1 | 4 - 5 | 311 | 133 | ||
| XM_011524813.1 | XP_011523115.1 | 4 - 5 | 638 | 133 | ||
| XM_017024642.1 | XP_016880131.1 | 5 - 6 | 1641 | 133 | ||
| XM_017024643.1 | XP_016880132.1 | 4 - 5 | 638 | 133 | ||
| XM_017024644.1 | XP_016880133.1 | 4 - 5 | 337 | 133 | ||
| GenBank mRNA | AK290005.1 | - | 4 - 5 | 285 | 133 | |
| AK297425.1 | - | 4 - 5 | 274 | 133 | ||
| AY450308.1 | - | 4 - 5 | 251 | 133 | ||
| BC009808.2 | - | 4 - 5 | 256 | 2989212 | 133 | |
| D30756.1 | - | 4 - 5 | 327 | 133 | ||
| DB081342.1 | - | 4 - 5 | 337 | 133 | ||
| X76952.1 | - | 4 - 5 | 279 | 133 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK297425.1 transcripts.
Set Membership: |
|

NBR1
Genomic Map