Assay Details
| Gene Symbol: CARD11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001324281.1 | NP_001311210.1 | 5 - 6 | 825 | 88 | |
| NM_032415.5 | NP_115791.3 | 4 - 5 | 764 | 88 | ||
| XM_011515586.2 | XP_011513888.1 | 5 - 6 | 793 | 88 | ||
| XM_011515587.2 | XP_011513889.1 | 4 - 5 | 759 | 88 | ||
| GenBank mRNA | AF322641.1 | - | 4 - 5 | 666 | 88 | |
| AF352576.1 | - | 2 - 3 | 339 | 88 | ||
| AK074049.1 | - | 4 - 5 | 684 | 88 | ||
| BC111719.1 | - | 3 - 4 | 450 | 40009168 | 88 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001102.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of cytokine production
stimulatory C-type lectin receptor signaling pathway
positive regulation of B cell proliferation
T cell costimulation
Fc-epsilon receptor signaling pathway
positive regulation of T cell proliferation
regulation of apoptotic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
thymic T cell selection
positive regulation of interleukin-2 biosynthetic process
regulation of B cell differentiation
regulation of T cell differentiation
GMP metabolic process
GDP metabolic process
T cell receptor signaling pathway
positive regulation of NF-kappaB transcription factor activity
guanylate kinase activity
protein binding
CARD domain binding
positive regulation of cytokine production
stimulatory C-type lectin receptor signaling pathway
positive regulation of B cell proliferation
T cell costimulation
Fc-epsilon receptor signaling pathway
positive regulation of T cell proliferation
regulation of apoptotic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
thymic T cell selection
positive regulation of interleukin-2 biosynthetic process
regulation of B cell differentiation
regulation of T cell differentiation
GMP metabolic process
GDP metabolic process
T cell receptor signaling pathway
positive regulation of NF-kappaB transcription factor activity
guanylate kinase activity
protein binding
CARD domain binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07017017.hg.1 | TC07001102.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07017017.hg.1 | ENST00000396946.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07017017.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006577.hg.1 | ENST00000423194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006577.hg.1 | ENST00000423194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006577.hg.1 | ENST00000423194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006577.hg.1 | ENST00000423194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000355508.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000355508.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000355508.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000355508.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000355508.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000355508.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000355508.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000356408.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000356408.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000356408.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000356408.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000356408.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000480332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000480332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000480332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000480332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000480332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | ENST00000480332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000355508.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000355508.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000355508.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000355508.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000355508.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000355508.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000355508.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000356408.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000356408.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000356408.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000356408.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000356408.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000480332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000480332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000480332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000480332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000480332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | ENST00000480332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010081.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700171818.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700171818.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700171818.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700171818.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700171818.hg.1 | TC0700010081.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700006577.hg.1 | ENST00000423194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006577.hg.1 | ENST00000423194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006577.hg.1 | ENST00000423194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006577.hg.1 | ENST00000423194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700171817.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700171817.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700171817.hg.1 | ENST00000396946.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700171817.hg.1 | NM_032415.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700171817.hg.1 | TC0700010081.hg.1 | Clariom D TC |

CARD11
Genomic Map