Assay Details
| Gene Symbol: BYSL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004053.3 | NP_004044.3 | 4 - 5 | 1041 | 62 | |
| GenBank mRNA | AK095253.1 | - | 1461 | 62 | ||
| AK314147.1 | - | 4 - 5 | 796 | 62 | ||
| BC007340.2 | - | 4 - 5 | 777 | 2989807 | 62 | |
| BC050645.1 | - | 4 - 5 | 1041 | 6011459 | 62 | |
| BC062627.1 | - | 4 - 5 | 1012 | 5226210 | 62 | |
| L36720.2 | - | 3 - 4 | 321 | 62 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000577.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
maturation of SSU-rRNA from tricistronic rRNA transcript (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
trophectodermal cell differentiation
rRNA processing
cell adhesion
brain development
female pregnancy
cell proliferation
neuron differentiation
cellular response to lipopolysaccharide
cellular response to interleukin-1
cellular response to growth factor stimulus
cellular response to organic cyclic compound
protein binding
poly(A) RNA binding
maturation of SSU-rRNA from tricistronic rRNA transcript (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
trophectodermal cell differentiation
rRNA processing
cell adhesion
brain development
female pregnancy
cell proliferation
neuron differentiation
cellular response to lipopolysaccharide
cellular response to interleukin-1
cellular response to growth factor stimulus
cellular response to organic cyclic compound
protein binding
poly(A) RNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06006491.hg.1 | TC06000577.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06006491.hg.1 | ENST00000230340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06006491.hg.1 | NM_004053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | ENST00000230340.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | ENST00000230340.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | ENST00000230340.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | ENST00000230340.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | ENST00000230340.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | ENST00000230340.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | ENST00000230340.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | ENST00000230340.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | ENST00000230340.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | ENST00000230340.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | ENST00000230340.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | ENST00000230340.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | ENST00000230340.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | ENST00000230340.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | ENST00000230340.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | ENST00000372996.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | ENST00000372996.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | ENST00000372996.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | ENST00000372996.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | ENST00000372996.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | ENST00000372996.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | ENST00000372996.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | ENST00000372996.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | ENST00000475702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | ENST00000475702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | ENST00000475702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | ENST00000475702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | ENST00000475702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | ENST00000489290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | ENST00000489290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | ENST00000489290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | ENST00000489290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | ENST00000489290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | ENST00000489290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | ENST00000489290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | ENST00000494032.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | ENST00000494032.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | ENST00000494032.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | ENST00000494032.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | NM_004053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | NM_004053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | NM_004053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | NM_004053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | NM_004053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | NM_004053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | NM_004053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | NM_004053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | NM_004053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | NM_004053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | NM_004053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | NM_004053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | NM_004053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008027.hg.1 | NM_004053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | ENST00000230340.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | ENST00000230340.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | ENST00000230340.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | ENST00000230340.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | ENST00000230340.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | ENST00000230340.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | ENST00000230340.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | ENST00000230340.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | ENST00000230340.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | ENST00000230340.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | ENST00000230340.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | ENST00000230340.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | ENST00000230340.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | ENST00000230340.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | ENST00000230340.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | ENST00000372996.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | ENST00000372996.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | ENST00000372996.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | ENST00000372996.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | ENST00000372996.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | ENST00000372996.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | ENST00000372996.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | ENST00000372996.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | ENST00000475702.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | ENST00000475702.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | ENST00000475702.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | ENST00000475702.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | ENST00000475702.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | ENST00000489290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | ENST00000489290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | ENST00000489290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | ENST00000489290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | ENST00000489290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | ENST00000489290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | ENST00000489290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | ENST00000494032.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | ENST00000494032.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | ENST00000494032.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | ENST00000494032.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | NM_004053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | NM_004053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | NM_004053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | NM_004053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | NM_004053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | NM_004053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | NM_004053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | NM_004053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | NM_004053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | NM_004053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | NM_004053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | NM_004053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | NM_004053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008027.hg.1 | NM_004053.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600155665.hg.1 | ENST00000230340.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600155665.hg.1 | ENST00000372996.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600155665.hg.1 | ENST00000489290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600155665.hg.1 | NM_004053.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600155665.hg.1 | ENST00000230340.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600155665.hg.1 | ENST00000372996.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600155665.hg.1 | ENST00000489290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600155665.hg.1 | NM_004053.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600155665.hg.1 | TC0600008027.hg.1 | Clariom D TC |

BYSL
Genomic Map