Assay Details
| Gene Symbol: KCNT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287819.1 | NP_001274748.1 | 21 - 22 | 2648 | 97 | |
| NM_001287820.1 | NP_001274749.1 | 20 - 21 | 2498 | 97 | ||
| NM_198503.3 | NP_940905.2 | 22 - 23 | 2720 | 97 | ||
| XM_006711294.2 | XP_006711357.1 | 22 - 23 | 3194 | 97 | ||
| XM_006711295.2 | XP_006711358.1 | 21 - 22 | 3122 | 97 | ||
| XM_011509482.2 | XP_011507784.1 | 21 - 22 | 3119 | 97 | ||
| XM_017001178.1 | XP_016856667.1 | 21 - 22 | 3122 | 97 | ||
| XM_017001181.1 | XP_016856670.1 | 12 - 13 | 1709 | 97 | ||
| XM_017001182.1 | XP_016856671.1 | 21 - 22 | 3085 | 97 | ||
| XM_017001183.1 | XP_016856672.1 | 9 - 10 | 1245 | 97 | ||
| XM_017001184.1 | XP_016856673.1 | 21 - 22 | 3085 | 97 | ||
| XM_017001185.1 | XP_016856674.1 | 20 - 21 | 3047 | 97 | ||
| GenBank mRNA | AK127807.1 | - | 14 - 15 | 1921 | 97 | |
| AK131498.1 | - | 20 - 21 | 2723 | 97 | ||
| AK309729.1 | - | 21 - 22 | 2637 | 97 | ||
| AY359444.1 | - | 22 - 23 | 2658 | 97 | ||
| BC103948.1 | - | 21 - 22 | 2576 | 40009146 | 97 | |
| BC103949.1 | - | 20 - 21 | 2426 | 40009147 | 97 | |
| BC103950.1 | - | 21 - 22 | 2576 | 40009148 | 97 | |
| BX647852.1 | - | 21 - 22 | 2630 | 97 | ||
| EU251076.1 | - | 22 - 23 | 2598 | 97 | ||
More Information
Set Membership: |
|

KCNT2
Genomic Map