Assay Details
| Gene Symbol: KCNT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287819.1 | NP_001274748.1 | 23 - 24 | 2828 | 77 | |
| NM_001287820.1 | NP_001274749.1 | 22 - 23 | 2678 | 77 | ||
| NM_198503.3 | NP_940905.2 | 24 - 25 | 2900 | 77 | ||
| XM_006711294.2 | XP_006711357.1 | 24 - 25 | 3374 | 77 | ||
| XM_006711295.2 | XP_006711358.1 | 23 - 24 | 3302 | 77 | ||
| XM_011509482.2 | XP_011507784.1 | 23 - 24 | 3299 | 77 | ||
| XM_017001178.1 | XP_016856667.1 | 23 - 24 | 3302 | 77 | ||
| XM_017001181.1 | XP_016856670.1 | 14 - 15 | 1889 | 77 | ||
| XM_017001182.1 | XP_016856671.1 | 23 - 24 | 3265 | 77 | ||
| XM_017001183.1 | XP_016856672.1 | 11 - 12 | 1425 | 77 | ||
| XM_017001184.1 | XP_016856673.1 | 23 - 24 | 3265 | 77 | ||
| XM_017001185.1 | XP_016856674.1 | 22 - 23 | 3227 | 77 | ||
| GenBank mRNA | AK127807.1 | - | 16 - 17 | 2101 | 77 | |
| AK131498.1 | - | 22 - 23 | 2903 | 77 | ||
| AK309729.1 | - | 23 - 24 | 2817 | 77 | ||
| AY359444.1 | - | 24 - 25 | 2838 | 77 | ||
| BC103947.1 | - | 18 - 19 | 2092 | 77 | ||
| BC103948.1 | - | 23 - 24 | 2756 | 40009146 | 77 | |
| BC103949.1 | - | 22 - 23 | 2606 | 40009147 | 77 | |
| BC103950.1 | - | 23 - 24 | 2756 | 40009148 | 77 | |
| BX647852.1 | - | 23 - 24 | 2810 | 77 | ||
| EU251076.1 | - | 24 - 25 | 2778 | 77 | ||
More Information
Set Membership: |
|

KCNT2
Genomic Map