Assay Details
| Gene Symbol: KCNT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287819.1 | NP_001274748.1 | 11 - 12 | 1244 | 88 | |
| NM_001287820.1 | NP_001274749.1 | 11 - 12 | 1244 | 88 | ||
| NM_198503.3 | NP_940905.2 | 11 - 12 | 1244 | 88 | ||
| XM_006711294.2 | XP_006711357.1 | 11 - 12 | 1718 | 88 | ||
| XM_006711295.2 | XP_006711358.1 | 11 - 12 | 1718 | 88 | ||
| XM_011509482.2 | XP_011507784.1 | 10 - 11 | 1643 | 88 | ||
| XM_011509483.2 | XP_011507785.1 | 11 - 12 | 1718 | 88 | ||
| XM_017001178.1 | XP_016856667.1 | 11 - 12 | 1718 | 88 | ||
| XM_017001179.1 | XP_016856668.1 | 11 - 12 | 1718 | 88 | ||
| XM_017001180.1 | XP_016856669.1 | 11 - 12 | 1718 | 88 | ||
| XM_017001182.1 | XP_016856671.1 | 11 - 12 | 1718 | 88 | ||
| XM_017001184.1 | XP_016856673.1 | 11 - 12 | 1718 | 88 | ||
| XM_017001185.1 | XP_016856674.1 | 10 - 11 | 1643 | 88 | ||
| GenBank mRNA | AK127807.1 | - | 4 - 5 | 594 | 88 | |
| AK131498.1 | - | 10 - 11 | 1298 | 88 | ||
| AK309729.1 | - | 11 - 12 | 1233 | 88 | ||
| AY359444.1 | - | 11 - 12 | 1182 | 88 | ||
| BC103948.1 | - | 11 - 12 | 1172 | 40009146 | 88 | |
| BC103949.1 | - | 11 - 12 | 1172 | 40009147 | 88 | |
| BC103950.1 | - | 11 - 12 | 1172 | 40009148 | 88 | |
| BX647852.1 | - | 11 - 12 | 1226 | 88 | ||
| EU251076.1 | - | 11 - 12 | 1122 | 88 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017001182.1, XM_017001184.1 transcripts.
Set Membership: |
|

KCNT2
Genomic Map