Assay Details
| Gene Symbol: PAPPA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020318.2 | NP_064714.2 | 14 - 15 | 5316 | 115 | |
| XM_005245422.3 | XP_005245479.1 | 14 - 15 | 5363 | 115 | ||
| XM_011509857.1 | XP_011508159.1 | 15 - 16 | 5233 | 115 | ||
| XM_011509858.2 | XP_011508160.1 | 14 - 15 | 5576 | 115 | ||
| XM_017002023.1 | XP_016857512.1 | 15 - 16 | 5414 | 115 | ||
| XM_017002024.1 | XP_016857513.1 | 15 - 16 | 5461 | 115 | ||
| XM_017002025.1 | XP_016857514.1 | 14 - 15 | 5567 | 115 | ||
| GenBank mRNA | AB209369.1 | - | 6 - 7 | 854 | 115 | |
| AF311940.1 | - | 13 - 14 | 4152 | 115 | ||
| AF342989.1 | - | 14 - 15 | 5316 | 115 | ||
| AJ278348.1 | - | 13 - 14 | 4115 | 115 | ||
| BC152552.1 | - | 14 - 15 | 5304 | 115 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001534.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01023673.hg.1 | TC01001534.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01023673.hg.1 | ENST00000367662.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01023673.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000479836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000479836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000479836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000479836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000486075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000486075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000486075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000493665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000493665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100178709.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100178709.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100178709.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100178709.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100178709.hg.1 | TC0100010713.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000479836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000479836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000479836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000479836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000486075.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000486075.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000486075.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000493665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000493665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_021936.2 | Refseq |

PAPPA2
Genomic Map