Assay Details
| Gene Symbol: N6AMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013240.5 | NP_037372.4 | 1 - 2 | 160 | 86 | |
| NM_182749.4 | NP_877426.4 | 1 - 2 | 160 | 86 | ||
| NR_047510.2 | 1 - 2 | 160 | 86 | |||
| GenBank mRNA | AF139682.1 | - | 1 - 2 | 161 | 86 | |
| AK225515.1 | - | 1 - 2 | 154 | 86 | ||
| AK314100.1 | - | 1 - 2 | 152 | 86 | ||
| BC011554.1 | - | 1 - 2 | 141 | 4640080 | 86 | |
| BP317563.1 | - | 1 - 2 | 157 | 86 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC21000350.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR21003967.hg.1 | ENST00000303775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003967.hg.1 | ENST00000351429.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003967.hg.1 | NM_013240.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003967.hg.1 | NM_182749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003967.hg.1 | TC21000350.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011978.hg.1 | ENST00000527217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011978.hg.1 | ENST00000527217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011978.hg.1 | ENST00000527217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011978.hg.1 | ENST00000527217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011978.hg.1 | ENST00000527217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003968.hg.1 | ENST00000303775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003968.hg.1 | ENST00000351429.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003968.hg.1 | NM_013240.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003968.hg.1 | NM_182749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003968.hg.1 | TC21000350.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | ENST00000303775.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | ENST00000303775.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | ENST00000303775.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | ENST00000303775.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | ENST00000303775.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | ENST00000303775.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | ENST00000303775.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | ENST00000303775.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | ENST00000303775.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | ENST00000303775.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | ENST00000303775.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | ENST00000351429.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | ENST00000351429.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | ENST00000351429.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | ENST00000351429.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | ENST00000351429.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | ENST00000351429.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | ENST00000351429.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | ENST00000351429.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | ENST00000351429.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | ENST00000351429.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | ENST00000460212.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | ENST00000460212.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | ENST00000460212.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | ENST00000460212.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | ENST00000460212.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | ENST00000460212.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | ENST00000460212.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | ENST00000460212.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | NM_013240.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | NM_013240.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | NM_013240.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | NM_013240.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | NM_013240.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | NM_013240.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | NM_013240.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | NM_013240.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | NM_013240.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | NM_182749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | NM_182749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | NM_182749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | NM_182749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | NM_182749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | NM_182749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | NM_182749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | NM_182749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | NR_047510.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | NR_047510.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | NR_047510.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | NR_047510.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | NR_047510.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | NR_047510.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | NR_047510.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007846.hg.1 | NR_047510.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | ENST00000303775.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | ENST00000303775.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | ENST00000303775.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | ENST00000303775.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | ENST00000303775.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | ENST00000303775.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | ENST00000303775.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | ENST00000303775.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | ENST00000303775.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | ENST00000303775.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | ENST00000303775.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | ENST00000351429.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | ENST00000351429.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | ENST00000351429.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | ENST00000351429.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | ENST00000351429.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | ENST00000351429.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | ENST00000351429.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | ENST00000351429.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | ENST00000351429.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | ENST00000351429.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | ENST00000460212.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | ENST00000460212.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | ENST00000460212.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | ENST00000460212.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | ENST00000460212.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | ENST00000460212.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | ENST00000460212.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | ENST00000460212.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | NM_013240.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | NM_013240.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | NM_013240.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | NM_013240.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | NM_013240.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | NM_013240.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | NM_013240.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | NM_013240.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | NM_013240.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | NM_182749.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | NM_182749.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | NM_182749.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | NM_182749.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | NM_182749.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | NM_182749.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | NM_182749.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | NM_182749.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | NR_047510.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | NR_047510.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | NR_047510.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | NR_047510.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | NR_047510.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | NR_047510.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | NR_047510.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007846.hg.1 | NR_047510.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011978.hg.1 | ENST00000527217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011978.hg.1 | ENST00000527217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011978.hg.1 | ENST00000527217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011978.hg.1 | ENST00000527217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011978.hg.1 | ENST00000527217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152890.hg.1 | ENST00000303775.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152890.hg.1 | ENST00000351429.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152890.hg.1 | ENST00000460212.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152890.hg.1 | NM_013240.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100152890.hg.1 | NM_182749.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100152890.hg.1 | NR_047510.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100152890.hg.1 | TC2100007846.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100152890.hg.1 | ENST00000303775.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152890.hg.1 | ENST00000351429.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152890.hg.1 | ENST00000460212.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152890.hg.1 | NM_013240.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100152890.hg.1 | NM_182749.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100152890.hg.1 | NR_047510.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100152889.hg.1 | ENST00000303775.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152889.hg.1 | ENST00000351429.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152889.hg.1 | ENST00000460212.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152889.hg.1 | NM_013240.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100152889.hg.1 | NM_182749.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100152889.hg.1 | NR_047510.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100152889.hg.1 | TC2100007846.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100152889.hg.1 | ENST00000303775.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152889.hg.1 | ENST00000351429.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152889.hg.1 | ENST00000460212.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152889.hg.1 | NM_013240.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100152889.hg.1 | NM_182749.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100152889.hg.1 | NR_047510.2 | Refseq |

N6AMT1
Genomic Map