Assay Details
| Gene Symbol: KDELR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100603.1 | NP_001094073.1 | 1 - 2 | 254 | 67 | |
| NM_006854.3 | NP_006845.1 | 1 - 2 | 254 | 67 | ||
| GenBank mRNA | AK315702.1 | - | 1 - 2 | 226 | 67 | |
| AL547016.3 | - | 1 - 2 | 231 | 67 | ||
| BC008081.2 | - | 1 - 2 | 158 | 2966703 | 67 | |
| BC012994.2 | - | 1 - 2 | 189 | 3604062 | 67 | |
| BC071982.1 | - | 1 - 2 | 208 | 5477638 | 67 | |
| DA713413.1 | - | 1 - 2 | 254 | 67 | ||
| M88458.1 | - | 1 - 2 | 131 | 67 | ||
| X63745.1 | - | 1 - 2 | 105 | 67 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07003350.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07017580.hg.1 | TC07003350.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07017580.hg.1 | ENST00000258739.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07017580.hg.1 | ENST00000490996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07017580.hg.1 | NM_001100603.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07017580.hg.1 | NM_006854.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | ENST00000258739.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | ENST00000258739.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | ENST00000258739.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | ENST00000258739.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | ENST00000258739.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | ENST00000258739.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | ENST00000258739.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | ENST00000258739.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | ENST00000258739.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | ENST00000258739.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | ENST00000258739.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | ENST00000258739.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | ENST00000258739.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | ENST00000258739.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | ENST00000382267.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | ENST00000382267.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | ENST00000382267.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | ENST00000382267.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | ENST00000382267.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | ENST00000454368.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | ENST00000454368.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | ENST00000454368.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | ENST00000454368.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | ENST00000454368.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | ENST00000454368.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | ENST00000454368.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | ENST00000454368.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | ENST00000454368.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | ENST00000454368.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | ENST00000462052.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | ENST00000462052.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | ENST00000462052.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | ENST00000462052.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | ENST00000490996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | ENST00000490996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | ENST00000490996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | ENST00000490996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | ENST00000490996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | ENST00000490996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | ENST00000490996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | NM_001100603.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | NM_001100603.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | NM_001100603.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | NM_001100603.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | NM_001100603.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | NM_001100603.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | NM_001100603.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | NM_001100603.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | NM_001100603.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | NM_001100603.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | NM_001100603.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | NM_001100603.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | NM_001100603.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | NM_006854.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | NM_006854.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | NM_006854.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | NM_006854.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | NM_006854.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | NM_006854.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | NM_006854.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | NM_006854.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | NM_006854.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | NM_006854.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | NM_006854.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | NM_006854.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | NM_006854.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013510.hg.1 | NM_006854.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07017582.hg.1 | TC07003350.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07017582.hg.1 | ENST00000258739.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07017582.hg.1 | ENST00000490996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07017582.hg.1 | NM_001100603.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07017582.hg.1 | NM_006854.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07017583.hg.1 | TC07003350.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07017583.hg.1 | ENST00000258739.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07017583.hg.1 | NM_001100603.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07017583.hg.1 | NM_006854.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07017589.hg.1 | TC07003350.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07017589.hg.1 | ENST00000258739.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07017589.hg.1 | NM_001100603.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07017589.hg.1 | NM_006854.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700199871.hg.1 | ENST00000258739.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700199871.hg.1 | ENST00000382267.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700199871.hg.1 | ENST00000462052.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700199871.hg.1 | ENST00000490996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700199871.hg.1 | NM_001100603.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700199871.hg.1 | NM_006854.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700199871.hg.1 | ENST00000258739.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700199871.hg.1 | ENST00000382267.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700199871.hg.1 | ENST00000462052.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700199871.hg.1 | ENST00000490996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700199871.hg.1 | NM_001100603.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700199871.hg.1 | NM_006854.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700199871.hg.1 | TC0700013510.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700199874.hg.1 | ENST00000258739.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700199874.hg.1 | ENST00000382267.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700199874.hg.1 | ENST00000462052.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700199874.hg.1 | NM_001100603.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700199874.hg.1 | NM_006854.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700199874.hg.1 | ENST00000258739.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700199874.hg.1 | ENST00000382267.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700199874.hg.1 | ENST00000462052.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700199874.hg.1 | NM_001100603.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700199874.hg.1 | NM_006854.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700199874.hg.1 | TC0700013510.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700199879.hg.1 | ENST00000258739.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700199879.hg.1 | NM_001100603.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700199879.hg.1 | NM_006854.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700199879.hg.1 | ENST00000258739.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700199879.hg.1 | NM_001100603.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700199879.hg.1 | NM_006854.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700199879.hg.1 | TC0700013510.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | ENST00000258739.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | ENST00000258739.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | ENST00000258739.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | ENST00000258739.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | ENST00000258739.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | ENST00000258739.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | ENST00000258739.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | ENST00000258739.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | ENST00000258739.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | ENST00000258739.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | ENST00000258739.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | ENST00000258739.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | ENST00000258739.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | ENST00000258739.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | ENST00000382267.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | ENST00000382267.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | ENST00000382267.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | ENST00000382267.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | ENST00000382267.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | ENST00000454368.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | ENST00000454368.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | ENST00000454368.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | ENST00000454368.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | ENST00000454368.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | ENST00000454368.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | ENST00000454368.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | ENST00000454368.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | ENST00000454368.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | ENST00000454368.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | ENST00000462052.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | ENST00000462052.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | ENST00000462052.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | ENST00000462052.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | ENST00000490996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | ENST00000490996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | ENST00000490996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | ENST00000490996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | ENST00000490996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | ENST00000490996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | ENST00000490996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | NM_001100603.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | NM_001100603.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | NM_001100603.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | NM_001100603.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | NM_001100603.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | NM_001100603.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | NM_001100603.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | NM_001100603.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | NM_001100603.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | NM_001100603.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | NM_001100603.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | NM_001100603.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | NM_001100603.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | NM_006854.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | NM_006854.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | NM_006854.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | NM_006854.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | NM_006854.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | NM_006854.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | NM_006854.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | NM_006854.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | NM_006854.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | NM_006854.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | NM_006854.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | NM_006854.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | NM_006854.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013510.hg.1 | NM_006854.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700199873.hg.1 | ENST00000258739.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700199873.hg.1 | ENST00000382267.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700199873.hg.1 | ENST00000462052.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700199873.hg.1 | ENST00000490996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700199873.hg.1 | NM_001100603.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700199873.hg.1 | NM_006854.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700199873.hg.1 | ENST00000258739.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700199873.hg.1 | ENST00000382267.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700199873.hg.1 | ENST00000462052.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700199873.hg.1 | ENST00000490996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700199873.hg.1 | NM_001100603.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700199873.hg.1 | NM_006854.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700199873.hg.1 | TC0700013510.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 159351 | - | - |

KDELR2
Genomic Map