Assay Details
| Gene Symbol: CNTFR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017014261.1 | XP_016869750.1 | 1 - 2 | 129 | 88 | |
| XM_017014263.1 | XP_016869752.1 | 1 - 2 | 158 | 88 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017014261.1, XM_017014263.1 transcripts.
Set Membership: |
|

CNTFR
Genomic Map