| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
ENST00000287600.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
ENST00000287600.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
ENST00000287600.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
ENST00000287600.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
ENST00000287600.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
ENST00000287600.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
ENST00000287600.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
ENST00000287600.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
ENST00000409772.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
ENST00000409772.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
ENST00000409772.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
ENST00000409772.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
ENST00000409772.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
ENST00000409772.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
ENST00000428104.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
ENST00000428104.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
ENST00000428104.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
ENST00000428104.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
ENST00000428104.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
ENST00000477748.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
ENST00000477748.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
ENST00000477748.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
ENST00000486044.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
ENST00000486044.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
ENST00000486044.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
ENST00000486044.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
ENST00000486044.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
NM_001291018.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
NM_001291018.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
NM_001291018.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
NM_001291018.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
NM_001291018.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
NM_001291018.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
NM_001291018.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
NM_002601.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
NM_002601.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
NM_002601.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
NM_002601.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
NM_002601.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
NM_002601.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
NM_002601.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
NM_002601.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
NR_110994.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
NR_110994.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016039.hg.1 |
NR_110994.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046227.hg.1 |
TC02002867.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046227.hg.1 |
ENST00000287600.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046227.hg.1 |
NM_002601.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200217855.hg.1 |
ENST00000287600.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200217855.hg.1 |
ENST00000409772.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200217855.hg.1 |
ENST00000428104.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200217855.hg.1 |
ENST00000486044.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200217855.hg.1 |
NM_001291018.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200217855.hg.1 |
NM_002601.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200217855.hg.1 |
ENST00000287600.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200217855.hg.1 |
ENST00000409772.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200217855.hg.1 |
ENST00000428104.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200217855.hg.1 |
ENST00000486044.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200217855.hg.1 |
NM_001291018.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200217855.hg.1 |
NM_002601.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200217855.hg.1 |
TC0200016039.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
ENST00000287600.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
ENST00000287600.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
ENST00000287600.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
ENST00000287600.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
ENST00000287600.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
ENST00000287600.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
ENST00000287600.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
ENST00000287600.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
ENST00000409772.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
ENST00000409772.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
ENST00000409772.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
ENST00000409772.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
ENST00000409772.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
ENST00000409772.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
ENST00000428104.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
ENST00000428104.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
ENST00000428104.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
ENST00000428104.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
ENST00000428104.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
ENST00000477748.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
ENST00000477748.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
ENST00000477748.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
ENST00000486044.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
ENST00000486044.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
ENST00000486044.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
ENST00000486044.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
ENST00000486044.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
NM_001291018.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
NM_001291018.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
NM_001291018.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
NM_001291018.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
NM_001291018.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
NM_001291018.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
NM_001291018.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
NM_002601.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
NM_002601.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
NM_002601.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
NM_002601.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
NM_002601.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
NM_002601.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
NM_002601.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
NM_002601.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
NR_110994.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
NR_110994.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016039.hg.1 |
NR_110994.1 |
Refseq |