Assay Details
| Gene Symbol: SNX24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017009394.1 | XP_016864883.1 | 1 - 2 | 56 | 116 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017009394.1 transcripts.
Set Membership: |
|

SNX24
Genomic Map