Assay Details
| Gene Symbol: BEND5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011542141.2 | XP_011540443.1 | 1 - 2 | 2863 | 85 | |
| XM_017002332.1 | XP_016857821.1 | 2 - 3 | 273 | 85 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011542141.2, XM_017002332.1 transcripts.
Set Membership: |
|

BEND5
Genomic Map