Assay Details
| Gene Symbol: SLC8A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006712085.3 | XP_006712148.1 | 1 - 2 | 1003 | 99 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006712085.3 transcripts.
Set Membership: |
|

SLC8A1
Genomic Map