Assay Details
| Gene Symbol: PCSK6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291309.1 | NP_001278238.1 | 13 - 14 | 2169 | 67 | |
| NM_002570.4 | NP_002561.1 | 15 - 16 | 2391 | 67 | ||
| GenBank mRNA | AB208955.1 | - | 12 - 13 | 1570 | 67 | |
| BX428035.2 | - | 6 - 7 | 709 | 67 | ||
| D87993.1 | - | 15 - 16 | 2151 | 67 | ||
| M80482.1 | - | 15 - 16 | 2246 | 67 | ||
More Information
Set Membership: |
|

PCSK6
Genomic Map