Assay Details
| Gene Symbol: BEND5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302082.1 | NP_001289011.1 | 2 - 3 | 500 | 130 | |
| XM_017002334.1 | XP_016857823.1 | 3 - 4 | 547 | 130 | ||
| GenBank mRNA | AK021650.1 | - | 2 - 3 | 362 | 130 | |
| KU178777.1 | - | 2 - 3 | 362 | 130 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK021650.1,NM_001302082.1,XM_017002334.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | ENST00000371833.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | ENST00000371833.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | ENST00000371833.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | ENST00000371833.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | ENST00000371833.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | ENST00000371833.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | ENST00000371833.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | ENST00000371833.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | ENST00000371833.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | ENST00000371833.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | ENST00000371833.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | ENST00000371833.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | ENST00000463562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | ENST00000463562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | ENST00000463562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | ENST00000463562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | ENST00000463562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | ENST00000463562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | ENST00000463562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | ENST00000463562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | ENST00000463562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | ENST00000463562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | ENST00000463562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | ENST00000463562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | ENST00000476079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | ENST00000476079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | ENST00000476079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | ENST00000476096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | ENST00000476096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | ENST00000476096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | ENST00000476096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | ENST00000480399.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | ENST00000480399.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | ENST00000480399.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | ENST00000480399.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | ENST00000480399.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | ENST00000480399.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | ENST00000480399.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | ENST00000489956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | ENST00000489956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | ENST00000489956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | NM_001302082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | NM_001302082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | NM_001302082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | NM_001302082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | NM_001302082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | NM_001302082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | NM_001302082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | NM_001302082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | NM_001302082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | NM_001302082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | NM_001302082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | NM_001302082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | NM_001302082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | NM_001302082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | NM_024603.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | NM_024603.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | NM_024603.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | NM_024603.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | NM_024603.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | NM_024603.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | NM_024603.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | NM_024603.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | NM_024603.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | NM_024603.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | NM_024603.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | NM_024603.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014134.hg.1 | NM_024603.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100205751.hg.1 | ENST00000463562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100205751.hg.1 | ENST00000480399.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100205751.hg.1 | NM_001302082.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100205751.hg.1 | ENST00000463562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100205751.hg.1 | ENST00000480399.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100205751.hg.1 | NM_001302082.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100205751.hg.1 | TC0100014134.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | ENST00000371833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | ENST00000371833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | ENST00000371833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | ENST00000371833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | ENST00000371833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | ENST00000371833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | ENST00000371833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | ENST00000371833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | ENST00000371833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | ENST00000371833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | ENST00000371833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | ENST00000371833.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | ENST00000463562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | ENST00000463562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | ENST00000463562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | ENST00000463562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | ENST00000463562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | ENST00000463562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | ENST00000463562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | ENST00000463562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | ENST00000463562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | ENST00000463562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | ENST00000463562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | ENST00000463562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | ENST00000476079.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | ENST00000476079.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | ENST00000476079.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | ENST00000476096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | ENST00000476096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | ENST00000476096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | ENST00000476096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | ENST00000480399.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | ENST00000480399.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | ENST00000480399.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | ENST00000480399.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | ENST00000480399.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | ENST00000480399.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | ENST00000480399.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | ENST00000489956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | ENST00000489956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | ENST00000489956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | NM_001302082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | NM_001302082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | NM_001302082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | NM_001302082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | NM_001302082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | NM_001302082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | NM_001302082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | NM_001302082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | NM_001302082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | NM_001302082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | NM_001302082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | NM_001302082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | NM_001302082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | NM_001302082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | NM_024603.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | NM_024603.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | NM_024603.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | NM_024603.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | NM_024603.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | NM_024603.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | NM_024603.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | NM_024603.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | NM_024603.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | NM_024603.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | NM_024603.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | NM_024603.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014134.hg.1 | NM_024603.3 | Refseq |

BEND5
Genomic Map