Assay Details
| Gene Symbol: TUBG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320509.1 | NP_001307438.1 | 5 - 6 | 680 | 67 | |
| XM_017024457.1 | XP_016879946.1 | 5 - 6 | 481 | 67 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017024457.1 transcripts.
Set Membership: |
|

TUBG2
Genomic Map