Assay Details
| Gene Symbol: PAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019088.3 | NP_061961.2 | 2 - 3 | 409 | 69 | |
| NR_046384.1 | 2 - 3 | 409 | 69 | |||
| GenBank mRNA | AJ401156.1 | - | 2 - 3 | 234 | 69 | |
| AK001985.1 | - | 2 - 3 | 306 | 69 | ||
| AK294625.1 | - | 2 - 3 | 331 | 69 | ||
| BC000017.2 | - | 2 - 3 | 253 | 3504227 | 69 | |
| BC013402.1 | - | 2 - 3 | 325 | 2960015 | 69 | |
| DA821359.1 | - | 2 - 3 | 409 | 69 | ||
More Information
Set Membership: |
|

PAF1
Genomic Map