| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007092.hg.1 |
ENST00000259963.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007092.hg.1 |
ENST00000259963.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007092.hg.1 |
ENST00000259963.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007092.hg.1 |
ENST00000259963.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007092.hg.1 |
ENST00000259963.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007092.hg.1 |
ENST00000259963.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007092.hg.1 |
ENST00000259963.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007092.hg.1 |
ENST00000259963.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007092.hg.1 |
NM_016255.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007092.hg.1 |
NM_016255.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007092.hg.1 |
NM_016255.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007092.hg.1 |
NM_016255.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007092.hg.1 |
NM_016255.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007092.hg.1 |
NM_016255.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007092.hg.1 |
NM_016255.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007092.hg.1 |
NM_016255.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007092.hg.1 |
NM_016255.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001216.hg.1 |
TC06000115.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001216.hg.1 |
ENST00000259963.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001216.hg.1 |
ENST00000542476.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001216.hg.1 |
NM_016255.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007092.hg.1 |
ENST00000259963.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007092.hg.1 |
ENST00000259963.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007092.hg.1 |
ENST00000259963.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007092.hg.1 |
ENST00000259963.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007092.hg.1 |
ENST00000259963.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007092.hg.1 |
ENST00000259963.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007092.hg.1 |
ENST00000259963.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007092.hg.1 |
ENST00000259963.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007092.hg.1 |
NM_016255.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007092.hg.1 |
NM_016255.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007092.hg.1 |
NM_016255.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007092.hg.1 |
NM_016255.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007092.hg.1 |
NM_016255.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007092.hg.1 |
NM_016255.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007092.hg.1 |
NM_016255.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007092.hg.1 |
NM_016255.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007092.hg.1 |
NM_016255.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600148376.hg.1 |
ENST00000259963.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600148376.hg.1 |
NM_016255.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600148376.hg.1 |
ENST00000259963.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600148376.hg.1 |
NM_016255.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600148376.hg.1 |
TC0600007092.hg.1 |
Clariom D TC |