Assay Details
| Gene Symbol: OTUD7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK126002.1 | - | 5 - 6 | 502 | 141 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560598.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560598.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560598.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560598.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560598.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560598.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560598.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560598.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560598.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560598.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560598.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560598.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | NM_130901.2 | Refseq |

OTUD7A
Genomic Map