Assay Details
| Gene Symbol: TMCO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005245827.3 | XP_005245884.1 | 9 - 10 | 1015 | 105 | |
| XM_017000913.1 | XP_016856402.1 | 9 - 10 | 1023 | 105 | ||
| GenBank mRNA | AL049847.1 | - | 6 - 7 | 763 | 105 | |
| AX775877.1 | - | 8 - 9 | 1028 | 105 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000294543.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000294543.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000294543.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000294543.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000294543.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000294543.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000294543.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000294543.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000294543.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000294543.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000294543.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000294543.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000294543.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000294543.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000294543.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000294543.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000294543.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000294543.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000294543.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000294543.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000294543.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000294543.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000375127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000375127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000375127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000375127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000375127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000375127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000375127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000375127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000375127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000375127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000375127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000375127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000375127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000375127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000375127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000375127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000375127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000375127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000375127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000462171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000462171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000462171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000462171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000462171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000488449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000488449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000488449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000488449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000488449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000488449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000488449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000489135.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000489135.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000489135.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000489135.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000489135.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000489135.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000489135.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000489135.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000489135.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000489135.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000489135.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000489814.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000489814.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000489814.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000489814.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000489814.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000489814.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000489814.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000489814.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000489814.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000494342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000494342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000494342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000496528.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000496528.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000496528.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000496528.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000496528.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000496528.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000496528.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000496528.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | ENST00000496528.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | NM_181719.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | NM_181719.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | NM_181719.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | NM_181719.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | NM_181719.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | NM_181719.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | NM_181719.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | NM_181719.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | NM_181719.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | NM_181719.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | NM_181719.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | NM_181719.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | NM_181719.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | NM_181719.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | NM_181719.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | NM_181719.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | NM_181719.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | NM_181719.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | NM_181719.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | NM_181719.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | NM_181719.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013153.hg.1 | NM_181719.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000294543.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000294543.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000294543.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000294543.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000294543.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000294543.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000294543.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000294543.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000294543.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000294543.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000294543.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000294543.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000294543.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000294543.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000294543.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000294543.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000294543.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000294543.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000294543.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000294543.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000294543.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000294543.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000375127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000375127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000375127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000375127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000375127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000375127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000375127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000375127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000375127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000375127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000375127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000375127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000375127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000375127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000375127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000375127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000375127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000375127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000375127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000462171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000462171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000462171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000462171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000462171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000488449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000488449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000488449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000488449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000488449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000488449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000488449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000489135.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000489135.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000489135.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000489135.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000489135.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000489135.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000489135.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000489135.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000489135.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000489135.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000489135.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000489814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000489814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000489814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000489814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000489814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000489814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000489814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000489814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000489814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000494342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000494342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000494342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000496528.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000496528.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000496528.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000496528.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000496528.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000496528.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000496528.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000496528.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | ENST00000496528.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | NM_181719.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | NM_181719.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | NM_181719.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | NM_181719.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | NM_181719.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | NM_181719.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | NM_181719.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | NM_181719.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | NM_181719.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | NM_181719.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | NM_181719.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | NM_181719.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | NM_181719.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | NM_181719.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | NM_181719.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | NM_181719.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | NM_181719.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | NM_181719.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | NM_181719.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | NM_181719.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | NM_181719.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013153.hg.1 | NM_181719.5 | Refseq |

TMCO4
Genomic Map