Assay Details
| Gene Symbol: BTNL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159708.1 | NP_001153180.1 | 5 - 6 | 1173 | 101 | |
| NM_024850.2 | NP_079126.1 | 6 - 7 | 1194 | 122 | ||
| GenBank mRNA | AK025111.1 | - | 6 - 7 | 1112 | 122 | |
| AK225213.1 | - | 6 - 7 | 1112 | 122 | ||
More Information
Set Membership: |
|

BTNL8
Genomic Map