Assay Details
| Gene Symbol: WDR62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083961.1 | NP_001077430.1 | 21 - 22 | 2608 | 68 | |
| NM_173636.4 | NP_775907.4 | 21 - 22 | 2608 | 68 | ||
| XM_005258809.2 | XP_005258866.1 | 21 - 22 | 2608 | 68 | ||
| XM_011526837.1 | XP_011525139.1 | 21 - 22 | 2593 | 68 | ||
| XM_011526838.1 | XP_011525140.1 | 21 - 22 | 2608 | 68 | ||
| XM_011526839.1 | XP_011525141.1 | 19 - 20 | 2257 | 68 | ||
| XM_011526840.2 | XP_011525142.1 | 14 - 15 | 1603 | 68 | ||
| XM_011526841.2 | XP_011525143.1 | 12 - 13 | 1224 | 68 | ||
| XM_011526842.1 | XP_011525144.1 | 11 - 12 | 1056 | 68 | ||
| XM_011526843.1 | XP_011525145.1 | 4 - 5 | 383 | 68 | ||
| XM_011526844.2 | XP_011525146.1 | 4 - 5 | 380 | 68 | ||
| XM_017026665.1 | XP_016882154.1 | 21 - 22 | 2608 | 68 | ||
| GenBank mRNA | BC017261.2 | - | 3 - 4 | 317 | 68 | |
| BC058939.1 | - | 21 - 22 | 2552 | 68 | ||
| BQ880028.1 | - | 8 - 9 | 777 | 68 | ||
| BX647726.1 | - | 22 - 23 | 2669 | 68 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000493.hg
Additional Information:
This assay is in the 5'UTR region of the BC017261.2 transcripts.
Set Membership: |
|

WDR62
Genomic Map