Assay Details
| Gene Symbol: HNRNPK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318186.1 | NP_001305115.1 | 14 - 15 | 1347 | 91 | |
| NM_001318187.1 | NP_001305116.1 | 14 - 15 | 1347 | 91 | ||
| NM_001318188.1 | NP_001305117.1 | 15 - 16 | 1419 | 91 | ||
| NM_002140.4 | NP_002131.2 | 15 - 16 | 1580 | 91 | ||
| NM_031262.3 | NP_112552.1 | 15 - 16 | 1580 | 91 | ||
| NM_031263.3 | NP_112553.1 | 15 - 16 | 1419 | 91 | ||
| XM_005251960.2 | XP_005252017.1 | 15 - 16 | 1398 | 91 | ||
| XM_005251963.3 | XP_005252020.1 | 14 - 15 | 1360 | 91 | ||
| XM_005251965.2 | XP_005252022.1 | 14 - 15 | 1318 | 91 | ||
| XM_011518616.1 | XP_011516918.1 | 14 - 15 | 1360 | 91 | ||
| XM_017014668.1 | XP_016870157.1 | 15 - 16 | 1398 | 91 | ||
| XM_017014669.1 | XP_016870158.1 | 14 - 15 | 1318 | 91 | ||
| GenBank mRNA | AB209562.1 | - | 14 - 15 | 1276 | 91 | |
| AB451263.1 | - | 13 - 14 | 1193 | 91 | ||
| AB451390.1 | - | 13 - 14 | 1193 | 91 | ||
| AK096385.1 | - | 14 - 15 | 1205 | 91 | ||
| AK123117.1 | - | 9 - 10 | 2145 | 91 | ||
| AK291336.1 | - | 15 - 16 | 1402 | 91 | ||
| AK294067.1 | - | 14 - 15 | 1377 | 91 | ||
| AK300744.1 | - | 14 - 15 | 1290 | 91 | ||
| AK307962.1 | - | 16 - 17 | 1240 | 91 | ||
| AY911506.1 | - | 14 - 15 | 1256 | 91 | ||
| BC000355.2 | - | 14 - 15 | 1315 | 2964383 | 91 | |
| BC014980.1 | - | 15 - 16 | 1386 | 4906241 | 91 | |
| BC025321.1 | - | 15 - 16 | 1338 | 91 | ||
| CR456771.1 | - | 13 - 14 | 1193 | 91 | ||
| S74678.1 | - | 14 - 15 | 1300 | 91 | ||
| X72727.1 | - | 15 - 16 | 1402 | 91 | ||
More Information
Set Membership: |
|

HNRNPK
Genomic Map