Assay Details
| Gene Symbol: TSC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005272211.1 | XP_005272268.1 | 1 - 2 | 137 | 61 | |
| XM_017015097.1 | XP_016870586.1 | 1 - 2 | 303 | 61 | ||
| XM_017015098.1 | XP_016870587.1 | 1 - 2 | 137 | 61 | ||
| XM_017015100.1 | XP_016870589.1 | 1 - 2 | 305 | 61 | ||
| GenBank mRNA | AK299654.1 | - | 1 - 2 | 63 | 61 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK299654.1, XM_005272211.1, XM_017015097.1, XM_017015098.1, XM_017015100.1 transcripts.
Set Membership: |
|

TSC1
Genomic Map