Assay Details
| Gene Symbol: CUX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202543.1 | NP_001189472.1 | 3 - 4 | 354 | 88 | |
| NM_001202545.2 | NP_001189474.1 | 3 - 4 | 354 | 88 | ||
| NM_001202546.2 | NP_001189475.1 | 2 - 3 | 139 | 88 | ||
| NM_001913.4 | NP_001904.2 | 3 - 4 | 354 | 88 | ||
| NM_181500.3 | NP_852477.1 | 3 - 4 | 354 | 88 | ||
| NM_181552.3 | NP_853530.2 | 3 - 4 | 232 | 88 | ||
| XM_005250150.2 | XP_005250207.1 | 3 - 4 | 753 | 88 | ||
| XM_005250151.2 | XP_005250208.1 | 3 - 4 | 756 | 88 | ||
| XM_005250154.3 | XP_005250211.1 | 3 - 4 | 610 | 88 | ||
| XM_006715854.2 | XP_006715917.1 | 3 - 4 | 753 | 88 | ||
| XM_006715855.1 | XP_006715918.1 | 3 - 4 | 610 | 88 | ||
| XM_011515823.2 | XP_011514125.1 | 3 - 4 | 751 | 88 | ||
| XM_011515825.1 | XP_011514127.1 | 3 - 4 | 319 | 88 | ||
| XM_017011759.1 | XP_016867248.1 | 3 - 4 | 319 | 88 | ||
| XM_017011760.1 | XP_016867249.1 | 3 - 4 | 319 | 88 | ||
| GenBank mRNA | AK122726.1 | - | 3 - 4 | 237 | 88 | |
| AK222832.1 | - | 3 - 4 | 250 | 88 | ||
| AK297548.1 | - | 3 - 4 | 240 | 88 | ||
| AK303151.1 | - | 2 - 3 | 143 | 88 | ||
| BC025422.2 | - | 3 - 4 | 235 | 88 | ||
| BC066592.1 | - | 3 - 4 | 247 | 5740343 | 88 | |
| L12579.1 | - | 3 - 4 | 246 | 88 | ||
| M74099.1 | - | 3 - 4 | 237 | 88 | ||
More Information
Set Membership: |
|

CUX1
Genomic Map