Assay Details
| Gene Symbol: LPCAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024830.3 | NP_079106.3 | 7 - 8 | 878 | 55 | |
| XM_005248373.3 | XP_005248430.1 | 8 - 9 | 786 | 55 | ||
| XM_011514132.1 | XP_011512434.1 | 7 - 8 | 887 | 55 | ||
| XM_011514134.1 | XP_011512436.1 | 7 - 8 | 653 | 55 | ||
| GenBank mRNA | AB244719.2 | - | 7 - 8 | 761 | 55 | |
| AK022499.1 | - | 2 - 3 | 57 | 55 | ||
| AK022505.1 | - | 2 - 3 | 57 | 55 | ||
| AK090444.1 | - | 5 - 6 | 357 | 55 | ||
| BC020166.1 | - | 5 - 6 | 344 | 55 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK022499.1, AK022505.1 transcripts.
Set Membership: |
|

LPCAT1
Genomic Map