Assay Details
| Gene Symbol: ARHGAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | U62794.1 | - | 5 - 6 | 590 | 58 | |
More Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000524594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000524594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000524594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000524594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000524594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000529960.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000529960.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000529960.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000529960.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000529960.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000529960.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000524594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000524594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000524594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000524594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000524594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000529960.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000529960.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000529960.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000529960.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000529960.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000529960.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |

ARHGAP1
Genomic Map