Assay Details
| Gene Symbol: CDHR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006718253.3 | XP_006718316.1 | 12 - 13 | 1448 | 87 | |
| XM_011520188.2 | XP_011518490.1 | 12 - 13 | 87 | |||
| XM_011520189.2 | XP_011518491.1 | 12 - 13 | 87 | |||
More Information
Set Membership: |
|

CDHR5
Genomic Map