Assay Details
Gene Symbol: GPR176 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011521167.2 | XP_011519469.1 | 2 - 3 | 582 | 75 | |
XM_017021877.1 | XP_016877366.1 | 2 - 3 | 565 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011521167.2, XM_017021877.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000299092.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000299092.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000299092.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000299092.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000299092.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000299092.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000299092.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000299092.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000299092.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000299092.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000543580.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000543580.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000543580.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000543580.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000543580.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000543580.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000543580.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000543580.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000543580.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000558041.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000558041.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000558041.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000558041.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000560729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000560729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000560729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000560729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000560729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000560729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000560729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000561100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000561100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000561100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000561100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000561100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000561100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000561100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000561100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000561100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000561100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_001271854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_001271854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_001271854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_001271854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_001271854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_001271854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_001271854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_001271854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_001271854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_001271854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_001271854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_001271855.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_001271855.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_001271855.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_001271855.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_001271855.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_001271855.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_001271855.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_001271855.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_001271855.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_007223.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_007223.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_007223.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_007223.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_007223.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_007223.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_007223.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_007223.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_007223.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_007223.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_007223.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000299092.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000299092.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000299092.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000299092.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000299092.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000299092.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000299092.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000299092.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000299092.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000299092.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000543580.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000543580.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000543580.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000543580.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000543580.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000543580.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000543580.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000543580.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000543580.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000558041.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000558041.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000558041.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000558041.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000560729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000560729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000560729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000560729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000560729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000560729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000560729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000561100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000561100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000561100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000561100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000561100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000561100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000561100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000561100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000561100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000561100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_001271854.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_001271854.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_001271854.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_001271854.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_001271854.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_001271854.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_001271854.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_001271854.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_001271854.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_001271854.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_001271854.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_001271855.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_001271855.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_001271855.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_001271855.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_001271855.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_001271855.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_001271855.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_001271855.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_001271855.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_007223.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_007223.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_007223.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_007223.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_007223.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_007223.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_007223.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_007223.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_007223.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_007223.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_007223.2 | Refseq |
