Assay Details
| Gene Symbol: AFG3L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006796.2 | NP_006787.2 | 6 - 7 | 820 | 73 | |
| XM_011525601.2 | XP_011523903.1 | 6 - 7 | 853 | 73 | ||
| GenBank mRNA | BC024282.1 | - | 7 - 8 | 673 | 73 | |
| BC065016.1 | - | 6 - 7 | 742 | 6169804 | 73 | |
| Y18314.1 | - | 6 - 7 | 739 | 73 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC18000373.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein complex assembly
axonogenesis
neuromuscular junction development
mitochondrial fusion
nerve development
mitochondrial protein processing
regulation of multicellular organism growth
cristae formation
myelination
protein import into mitochondrial intermembrane space
muscle fiber development
righting reflex
ATP-dependent peptidase activity
metalloendopeptidase activity
protein binding
ATP binding
metallopeptidase activity
zinc ion binding
unfolded protein binding
protein complex assembly
axonogenesis
neuromuscular junction development
mitochondrial fusion
nerve development
mitochondrial protein processing
regulation of multicellular organism growth
cristae formation
myelination
protein import into mitochondrial intermembrane space
muscle fiber development
righting reflex
ATP-dependent peptidase activity
metalloendopeptidase activity
protein binding
ATP binding
metallopeptidase activity
zinc ion binding
unfolded protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR18003884.hg.1 | ENST00000269143.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18003884.hg.1 | ENST00000537174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18003884.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18003884.hg.1 | TC18000373.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | ENST00000588893.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | ENST00000588893.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | ENST00000588893.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | ENST00000590811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | ENST00000590811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | ENST00000590811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | ENST00000590811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | ENST00000590811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | ENST00000590811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | ENST00000591848.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | ENST00000591848.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | ENST00000591848.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | ENST00000588893.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | ENST00000588893.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | ENST00000588893.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | ENST00000590811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | ENST00000590811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | ENST00000590811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | ENST00000590811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | ENST00000590811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | ENST00000590811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | ENST00000591848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | ENST00000591848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | ENST00000591848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008116.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800155492.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800155492.hg.1 | ENST00000590811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800155492.hg.1 | ENST00000591848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800155492.hg.1 | NM_006796.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800155492.hg.1 | TC1800008116.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800155492.hg.1 | ENST00000269143.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800155492.hg.1 | ENST00000590811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800155492.hg.1 | ENST00000591848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800155492.hg.1 | NM_006796.2 | Refseq |

AFG3L2
Genomic Map