Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020684.hg.1 |
TC06001731.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020684.hg.1 |
ENST00000053468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020684.hg.1 |
NM_018141.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011822.hg.1 |
ENST00000053468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011822.hg.1 |
ENST00000053468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011822.hg.1 |
ENST00000053468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011822.hg.1 |
ENST00000053468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011822.hg.1 |
ENST00000053468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011822.hg.1 |
ENST00000053468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011822.hg.1 |
ENST00000053468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011822.hg.1 |
ENST00000053468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011822.hg.1 |
ENST00000053468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011822.hg.1 |
NM_018141.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011822.hg.1 |
NM_018141.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011822.hg.1 |
NM_018141.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011822.hg.1 |
NM_018141.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011822.hg.1 |
NM_018141.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011822.hg.1 |
NM_018141.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011822.hg.1 |
NM_018141.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011822.hg.1 |
NM_018141.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011822.hg.1 |
NM_018141.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011822.hg.1 |
NM_018141.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011822.hg.1 |
ENST00000053468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011822.hg.1 |
ENST00000053468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011822.hg.1 |
ENST00000053468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011822.hg.1 |
ENST00000053468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011822.hg.1 |
ENST00000053468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011822.hg.1 |
ENST00000053468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011822.hg.1 |
ENST00000053468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011822.hg.1 |
ENST00000053468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011822.hg.1 |
ENST00000053468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011822.hg.1 |
NM_018141.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011822.hg.1 |
NM_018141.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011822.hg.1 |
NM_018141.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011822.hg.1 |
NM_018141.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011822.hg.1 |
NM_018141.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011822.hg.1 |
NM_018141.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011822.hg.1 |
NM_018141.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011822.hg.1 |
NM_018141.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011822.hg.1 |
NM_018141.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011822.hg.1 |
NM_018141.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600181204.hg.1 |
ENST00000053468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600181204.hg.1 |
NM_018141.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600181204.hg.1 |
ENST00000053468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600181204.hg.1 |
NM_018141.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600181204.hg.1 |
TC0600011822.hg.1 |
Clariom D TC |