Assay Details
| Gene Symbol: ANKRD66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017010148.1 | XP_016865637.1 | 5 | 2573 | 106 | |
| GenBank mRNA | AK057637.1 | - | 2 | 2228 | 106 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | phospholipase A2 group VII | PLA2G7 |
LDL-PLA2, LP-PLA2, PAFAD, PAFAH |
-20.4kb | Anti-sense | Chr. 6 - 46700954 - 46735836 on NCBI Build GRCh38 |
| Downstream | meprin A subunit alpha | MEP1A | PPHA | 34.9kb | Sense | Chr. 6 - 46793357 - 46845987 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

ANKRD66
Genomic Map