| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19015708.hg.1 |
ENST00000380951.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19015708.hg.1 |
ENST00000397910.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19015708.hg.1 |
NM_024690.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19015708.hg.1 |
TC19001139.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1900176999.hg.1 |
ENST00000397910.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900176999.hg.1 |
ENST00000596768.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900176999.hg.1 |
ENST00000599436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900176999.hg.1 |
ENST00000601404.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900176999.hg.1 |
NM_024690.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900176999.hg.1 |
TC1900009559.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900176999.hg.1 |
ENST00000397910.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900176999.hg.1 |
ENST00000596768.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900176999.hg.1 |
ENST00000599436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900176999.hg.1 |
ENST00000601404.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900176999.hg.1 |
NM_024690.2 |
Refseq |