Assay Details
| Gene Symbol: APPBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282476.1 | NP_001269405.1 | 6 - 7 | 1049 | 151 | |
| NM_006380.3 | NP_006371.2 | 7 - 8 | 1138 | 151 | ||
| XM_011524198.2 | XP_011522500.1 | 8 - 9 | 865 | 151 | ||
| XM_011524199.2 | XP_011522501.1 | 4 - 5 | 350 | 151 | ||
| GenBank mRNA | AF017782.1 | - | 7 - 8 | 1114 | 151 | |
| AK292227.1 | - | 7 - 8 | 905 | 151 | ||
| AK307666.1 | - | 6 - 7 | 943 | 151 | ||
| BC018121.1 | - | 7 - 8 | 1041 | 3916891 | 151 | |
| D86981.1 | - | 7 - 8 | 1117 | 151 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001764.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17023550.hg.1 | ENST00000083182.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17023550.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17023550.hg.1 | TC17001764.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000585368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000585368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000585368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000585368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000588668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000588668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000588668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000588668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000588668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000585368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000585368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000585368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000585368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000588668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000588668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000588668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000588668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000588668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700191247.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700191247.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700191247.hg.1 | ENST00000592995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700191247.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700191247.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700191247.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700191247.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700191247.hg.1 | ENST00000592995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700191247.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700191247.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700191247.hg.1 | TC1700011319.hg.1 | Clariom D TC |

APPBP2
Genomic Map