Assay Details
| Gene Symbol: CACNA2D4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK301234.1 | - | 1 - 2 | 135 | 96 | |
| BC048288.1 | - | 1 - 2 | 71 | 96 | ||
| BI908421.1 | - | 1 - 2 | 101 | 96 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK301234.1, BC048288.1 transcripts.
Set Membership: |
|

CACNA2D4
Genomic Map