Assay Details
| Gene Symbol: CD79B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK222954.1 | - | 2 - 3 | 185 | 70 | |
| AK223210.1 | - | 2 - 3 | 190 | 70 | ||
| AK313583.1 | - | 2 - 3 | 182 | 70 | ||
| AX775815.1 | - | 2 - 3 | 181 | 70 | ||
| AX775817.1 | - | 2 - 3 | 178 | 70 | ||
| BC002975.1 | - | 2 - 3 | 165 | 3542354 | 70 | |
| BC032651.1 | - | 2 - 3 | 148 | 5504755 | 70 | |
| M80461.1 | - | 2 - 3 | 128 | 70 | ||
| M89957.1 | - | 2 - 3 | 206 | 70 | ||
| S52229.1 | - | 2 - 3 | 112 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | ENST00000006750.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | ENST00000006750.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | ENST00000006750.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | ENST00000006750.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | ENST00000006750.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | ENST00000006750.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | ENST00000006750.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | ENST00000006750.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | ENST00000006750.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | ENST00000349817.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | ENST00000349817.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | ENST00000349817.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | ENST00000349817.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | ENST00000349817.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | ENST00000392795.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | ENST00000392795.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | ENST00000392795.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | ENST00000392795.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | ENST00000392795.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | ENST00000392795.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | ENST00000392795.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | ENST00000392795.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | ENST00000392795.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | ENST00000558969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | ENST00000558969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | ENST00000558969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | ENST00000558969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | ENST00000559358.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | ENST00000559358.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | ENST00000559358.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | ENST00000559358.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | ENST00000559358.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | ENST00000559358.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | ENST00000583260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | ENST00000583260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | ENST00000583260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | NM_000626.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | NM_000626.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | NM_000626.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | NM_000626.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | NM_000626.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | NM_000626.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | NM_000626.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | NM_000626.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | NM_000626.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | NM_001039933.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | NM_001039933.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | NM_001039933.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | NM_001039933.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | NM_001039933.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | NM_001039933.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | NM_001039933.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | NM_001039933.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | NM_001039933.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | NM_021602.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | NM_021602.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | NM_021602.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | NM_021602.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | NM_021602.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | NM_021602.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | NM_021602.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011430.hg.1 | NM_021602.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | ENST00000006750.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | ENST00000006750.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | ENST00000006750.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | ENST00000006750.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | ENST00000006750.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | ENST00000006750.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | ENST00000006750.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | ENST00000006750.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | ENST00000006750.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | ENST00000349817.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | ENST00000349817.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | ENST00000349817.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | ENST00000349817.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | ENST00000349817.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | ENST00000392795.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | ENST00000392795.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | ENST00000392795.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | ENST00000392795.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | ENST00000392795.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | ENST00000392795.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | ENST00000392795.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | ENST00000392795.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | ENST00000392795.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | ENST00000558969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | ENST00000558969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | ENST00000558969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | ENST00000558969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | ENST00000559358.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | ENST00000559358.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | ENST00000559358.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | ENST00000559358.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | ENST00000559358.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | ENST00000559358.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | ENST00000583260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | ENST00000583260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | ENST00000583260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | NM_000626.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | NM_000626.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | NM_000626.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | NM_000626.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | NM_000626.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | NM_000626.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | NM_000626.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | NM_000626.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | NM_000626.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | NM_001039933.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | NM_001039933.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | NM_001039933.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | NM_001039933.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | NM_001039933.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | NM_001039933.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | NM_001039933.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | NM_001039933.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | NM_001039933.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | NM_021602.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | NM_021602.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | NM_021602.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | NM_021602.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | NM_021602.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | NM_021602.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | NM_021602.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011430.hg.1 | NM_021602.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700192228.hg.1 | ENST00000006750.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700192228.hg.1 | ENST00000392795.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700192228.hg.1 | ENST00000559358.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700192228.hg.1 | NM_000626.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700192228.hg.1 | NM_001039933.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700192228.hg.1 | ENST00000006750.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700192228.hg.1 | ENST00000392795.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700192228.hg.1 | ENST00000559358.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700192228.hg.1 | NM_000626.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700192228.hg.1 | NM_001039933.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700192228.hg.1 | TC1700011430.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 119574 | - | - |

CD79B
Genomic Map