Assay Details
| Gene Symbol: TBCC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC017479.1 | - | 1 | 241 | 3357283 | 79 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC017479.1 transcripts.
Set Membership: |
|

TBCC
Genomic Map