Assay Details
| Gene Symbol: CIR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004882.3 | NP_004873.3 | 1 - 2 | 168 | 148 | |
| GenBank mRNA | AF098297.1 | - | 1 - 2 | 75 | 148 | |
| AK292389.1 | - | 1 - 2 | 151 | 148 | ||
| BC015040.1 | - | 1 - 2 | 122 | 148 | ||
| BC021175.1 | - | 1 - 2 | 136 | 148 | ||
| BC031018.1 | - | 1 - 2 | 99 | 148 | ||
| BC038987.1 | - | 1 - 2 | 143 | 148 | ||
| BC046098.2 | - | 1 - 2 | 119 | 5495320 | 148 | |
| BU596189.1 | - | 1 - 2 | 171 | 148 | ||
| U03644.1 | - | 1 - 2 | 107 | 148 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02002538.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
mRNA processing
RNA splicing
negative regulation of transcription, DNA-templated
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
transcription corepressor activity
protein binding
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
mRNA processing
RNA splicing
negative regulation of transcription, DNA-templated
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
transcription corepressor activity
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400011658.hg.1 | ENST00000514821.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | ENST00000342016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | ENST00000342016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | ENST00000342016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | ENST00000342016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | ENST00000342016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | ENST00000342016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | ENST00000342016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | ENST00000342016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | ENST00000342016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | ENST00000342016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | ENST00000342016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | ENST00000342016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | ENST00000342016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | ENST00000342016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | ENST00000342016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | ENST00000342016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | ENST00000377973.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | ENST00000377973.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | ENST00000377973.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | ENST00000377973.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | ENST00000461454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | ENST00000461454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | ENST00000461454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | ENST00000461454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | ENST00000461454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | ENST00000461454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | ENST00000461454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | ENST00000461454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | ENST00000461454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | ENST00000461454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | ENST00000464393.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | ENST00000464393.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | ENST00000464393.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | ENST00000464393.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | ENST00000472169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | ENST00000472169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | ENST00000472169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | ENST00000472169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | ENST00000472169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | NM_004882.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | NM_004882.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | NM_004882.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | NM_004882.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | NM_004882.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | NM_004882.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | NM_004882.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | NM_004882.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | NM_004882.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | NM_004882.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | NM_004882.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | NM_004882.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | NM_004882.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | NM_004882.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014990.hg.1 | NM_004882.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02040259.hg.1 | TC02002538.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02040259.hg.1 | ENST00000342016.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02040259.hg.1 | NM_004882.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012555.hg.1 | ENST00000524055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02040258.hg.1 | TC02002538.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02040258.hg.1 | ENST00000342016.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02040258.hg.1 | NM_004882.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007256.hg.1 | ENST00000405137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007256.hg.1 | ENST00000405137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200209348.hg.1 | ENST00000342016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200209348.hg.1 | ENST00000461454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200209348.hg.1 | ENST00000472169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200209348.hg.1 | NM_004882.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200209348.hg.1 | ENST00000342016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200209348.hg.1 | ENST00000461454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200209348.hg.1 | ENST00000472169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200209348.hg.1 | NM_004882.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200209348.hg.1 | TC0200014990.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400011658.hg.1 | ENST00000514821.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012555.hg.1 | ENST00000524055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007256.hg.1 | ENST00000405137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007256.hg.1 | ENST00000405137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200209346.hg.1 | ENST00000342016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200209346.hg.1 | ENST00000461454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200209346.hg.1 | ENST00000472169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200209346.hg.1 | NM_004882.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200209346.hg.1 | ENST00000342016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200209346.hg.1 | ENST00000461454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200209346.hg.1 | ENST00000472169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200209346.hg.1 | NM_004882.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200209346.hg.1 | TC0200014990.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200209347.hg.1 | ENST00000342016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200209347.hg.1 | ENST00000461454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200209347.hg.1 | ENST00000472169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200209347.hg.1 | NM_004882.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200209347.hg.1 | ENST00000342016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200209347.hg.1 | ENST00000461454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200209347.hg.1 | ENST00000472169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200209347.hg.1 | NM_004882.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200209347.hg.1 | TC0200014990.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200209349.hg.1 | ENST00000342016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200209349.hg.1 | ENST00000461454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200209349.hg.1 | ENST00000472169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200209349.hg.1 | NM_004882.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200209349.hg.1 | ENST00000342016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200209349.hg.1 | ENST00000461454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200209349.hg.1 | ENST00000472169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200209349.hg.1 | NM_004882.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200209349.hg.1 | TC0200014990.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | ENST00000342016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | ENST00000342016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | ENST00000342016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | ENST00000342016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | ENST00000342016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | ENST00000342016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | ENST00000342016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | ENST00000342016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | ENST00000342016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | ENST00000342016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | ENST00000342016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | ENST00000342016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | ENST00000342016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | ENST00000342016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | ENST00000342016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | ENST00000342016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | ENST00000377973.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | ENST00000377973.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | ENST00000377973.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | ENST00000377973.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | ENST00000461454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | ENST00000461454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | ENST00000461454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | ENST00000461454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | ENST00000461454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | ENST00000461454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | ENST00000461454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | ENST00000461454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | ENST00000461454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | ENST00000461454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | ENST00000464393.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | ENST00000464393.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | ENST00000464393.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | ENST00000464393.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | ENST00000472169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | ENST00000472169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | ENST00000472169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | ENST00000472169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | ENST00000472169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | NM_004882.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | NM_004882.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | NM_004882.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | NM_004882.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | NM_004882.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | NM_004882.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | NM_004882.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | NM_004882.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | NM_004882.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | NM_004882.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | NM_004882.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | NM_004882.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | NM_004882.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | NM_004882.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014990.hg.1 | NM_004882.3 | Refseq |

CIR1
Genomic Map