Assay Details
| Gene Symbol: RPL30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000989.3 | NP_000980.1 | 4 - 5 | 412 | 103 | |
| GenBank mRNA | BC032700.2 | - | 3 - 4 | 340 | 5393199 | 103 |
| BC095426.1 | - | 4 - 5 | 361 | 30792764 | 103 | |
| BM785413.1 | - | 4 - 5 | 412 | 103 | ||
| BQ055546.1 | - | 3 - 4 | 314 | 103 | ||
| L05095.1 | - | 4 - 5 | 351 | 103 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08001458.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
rRNA processing
translation
translational initiation
SRP-dependent cotranslational protein targeting to membrane
viral transcription
RNA binding
structural constituent of ribosome
protein binding
poly(A) RNA binding
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
rRNA processing
translation
translational initiation
SRP-dependent cotranslational protein targeting to membrane
viral transcription
RNA binding
structural constituent of ribosome
protein binding
poly(A) RNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR08018861.hg.1 | TC08001458.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08018861.hg.1 | ENST00000287038.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08018861.hg.1 | ENST00000396070.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08018861.hg.1 | ENST00000521291.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08018861.hg.1 | NM_000989.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000287038.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000287038.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000287038.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000287038.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000287038.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000287038.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000287038.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000396070.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000396070.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000396070.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000396070.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000396070.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000517489.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000517489.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000517489.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000517489.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000517489.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000517805.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000517805.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000517805.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000518164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000518164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000518164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000518850.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000518850.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000518850.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000518850.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000518850.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000518850.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000518850.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000518850.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000520998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000520998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000520998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000520998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000520998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000521112.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000521112.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000521112.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000521112.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000521291.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000521291.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000521291.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000521291.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000521291.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000521291.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000521534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000521534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000521534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000521534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000521534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000521534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000521726.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000521726.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000521726.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000521726.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000521726.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000522156.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000522156.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000522156.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000522156.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000523172.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000523172.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000523172.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000523172.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | NM_000989.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | NM_000989.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | NM_000989.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | NM_000989.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | NM_000989.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | NM_000989.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | NM_000989.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800176222.hg.1 | ENST00000287038.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800176222.hg.1 | ENST00000396070.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800176222.hg.1 | ENST00000518850.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800176222.hg.1 | ENST00000521291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800176222.hg.1 | ENST00000521534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800176222.hg.1 | NM_000989.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800176222.hg.1 | ENST00000287038.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800176222.hg.1 | ENST00000396070.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800176222.hg.1 | ENST00000518850.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800176222.hg.1 | ENST00000521291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800176222.hg.1 | ENST00000521534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800176222.hg.1 | NM_000989.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800176222.hg.1 | TC0800011206.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000287038.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000287038.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000287038.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000287038.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000287038.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000287038.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000287038.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000396070.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000396070.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000396070.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000396070.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000396070.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000517489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000517489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000517489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000517489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000517489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000517805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000517805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000517805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000518164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000518164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000518164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000518850.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000518850.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000518850.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000518850.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000518850.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000518850.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000518850.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000518850.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000520998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000520998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000520998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000520998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000520998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000521112.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000521112.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000521112.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000521112.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000521291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000521291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000521291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000521291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000521291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000521291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000521534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000521534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000521534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000521534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000521534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000521534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000521726.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000521726.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000521726.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000521726.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000521726.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000522156.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000522156.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000522156.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000522156.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000523172.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000523172.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000523172.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000523172.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | NM_000989.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | NM_000989.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | NM_000989.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | NM_000989.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | NM_000989.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | NM_000989.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | NM_000989.3 | Refseq |

RPL30
Genomic Map