Assay Details
| Gene Symbol: TMEM267 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005248333.4 | XP_005248390.1 | 1 - 2 | 843 | 97 | |
| XM_011514074.2 | XP_011512376.1 | 1 - 2 | 843 | 97 | ||
| XM_011514075.2 | XP_011512377.1 | 1 - 2 | 843 | 97 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005248333.4, XM_011514074.2, XM_011514075.2 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500007298.hg.1 | ENST00000504469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007298.hg.1 | ENST00000504469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007298.hg.1 | ENST00000504469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007298.hg.1 | ENST00000505645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007298.hg.1 | ENST00000505645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007298.hg.1 | ENST00000505645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000511525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000511525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000511525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000511525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007298.hg.1 | ENST00000504469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007298.hg.1 | ENST00000504469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007298.hg.1 | ENST00000504469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007298.hg.1 | ENST00000505645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007298.hg.1 | ENST00000505645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007298.hg.1 | ENST00000505645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000511525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000511525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000511525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000511525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |

TMEM267
Genomic Map