Assay Details
| Gene Symbol: UBAC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006719948.3 | XP_006720011.1 | 1 - 2 | 82 | 95 | |
| XM_011521083.2 | XP_011519385.1 | 1 - 2 | 79 | 95 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006719948.3, XM_011521083.2 transcripts.
Set Membership: |
|

UBAC2
Genomic Map