Assay Details
| Gene Symbol: UBAC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK124404.1 | - | 6 - 7 | 460 | 76 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK124404.1 transcripts.
Set Membership: |
|

UBAC2
Genomic Map